missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SENP6 Antibody (4B7), Novus Biologicals™

Product Code. 18346909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346909 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346909 Supplier Novus Biologicals™ Supplier No. H00026054M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SENP6 Monoclonal antibody specifically detects SENP6 in Human,Mouse samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Sandwich ELISA,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP6
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Sandwich ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 4B7
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Sandwich ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_056386
Gene Alias EC 3.4.22.-, KIAA0797FLJ11887, Sentrin/SUMO-specific protease SENP6, sentrin-specific protease 6, SSP1KIAA0389, SUMO1/sentrin specific peptidase 6,2810017C20Rik, SUMO1/sentrin specific protease 6, SUMO-1-specific protease 1, SUSP1FLJ11355
Host Species Mouse
Immunogen SENP6 (NP_056386, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAGKSGGSAGEITFLEALARSESKRDGGFKNNWSFDHEEESEGDTDKDGTNLLSVDEDEDSETSKGKKLNRRSEIVANSSGEFILKTYVRRNKSESFKTLKGNPIGLNM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26054
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.