missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £486.00
Specifications
| Antigen | SENP3 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18406711
|
Novus Biologicals
NBP2-32525-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018453
|
Novus Biologicals
NBP2-32525 |
0.1 mL |
£486.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SENP3 Polyclonal specifically detects SENP3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SENP3 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 26168 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LREEHVTCVQSILDEFLQTYGSLIPLSTDEVVEKLEDIFQQEFSTPSRKGLVLQLIQSYQRMPGNAMVRGFRVAYKRHVLTMDDLGTL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZP586K0919, DKFZp762A152, EC 3.4.22, EC 3.4.22.-, sentrin/SUMO-specific protease 3, Sentrin/SUMO-specific protease SENP3, sentrin-specific protease 3, SMT3IP1, SSP3DKFZp586K0919, SUMO1/sentrin/SMT3 specific peptidase 3, SUMO1/sentrin/SMT3 specific protease 3, SUMO-1-specific protease 3, SUSP3 | |
| SENP3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title