missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SENP3 Polyclonal antibody specifically detects SENP3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | SENP3 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZP586K0919, DKFZp762A152, EC 3.4.22, EC 3.4.22.-, sentrin/SUMO-specific protease 3, Sentrin/SUMO-specific protease SENP3, sentrin-specific protease 3, SMT3IP1, SSP3DKFZp586K0919, SUMO1/sentrin/SMT3 specific peptidase 3, SUMO1/sentrin/SMT3 specific protease 3, SUMO-1-specific protease 3, SUSP3 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 1 and 50 of human SUMO1/Sentrin/SMT3 Specific Peptidase 3 using the numbering given in entry NP_056485.2 (GeneID 26168). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?