missing translation for 'onlineSavingsMsg'
Learn More

SENP3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18424843 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424843 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424843 Supplier Novus Biologicals Supplier No. NBP171894AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SENP3 Polyclonal antibody specifically detects SENP3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias DKFZP586K0919, DKFZp762A152, EC 3.4.22, EC 3.4.22.-, sentrin/SUMO-specific protease 3, Sentrin/SUMO-specific protease SENP3, sentrin-specific protease 3, SMT3IP1, SSP3DKFZp586K0919, SUMO1/sentrin/SMT3 specific peptidase 3, SUMO1/sentrin/SMT3 specific protease 3, SUMO-1-specific protease 3, SUSP3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human SUMO1/Sentrin/SMT3 Specific Peptidase 3 using the numbering given in entry NP_056485.2 (GeneID 26168).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26168
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.