missing translation for 'onlineSavingsMsg'
Learn More

SENP2 Antibody (OTI3D3), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18433266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433266 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433266 Supplier Novus Biologicals Supplier No. NBP274062MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SENP2 Monoclonal antibody specifically detects SENP2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3D3
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 139-523 of human SENP2 (NP_067640) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59343
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.