missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | SENP2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18413941
|
Novus Biologicals
NBP1-88920-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18433941
|
Novus Biologicals
NBP1-88920 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SENP2 Polyclonal antibody specifically detects SENP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SENP2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RRGYQLEPDLSEEVSARLRLGSGSNGLLRRKVSIIETKEKNCSGKERDRRTDDLLELTEDMEKEISNALGHGPQDEILSSAFKL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 59343 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title