missing translation for 'onlineSavingsMsg'
Learn More

Senataxin Antibody (BLR050F), Novus Biologicals™

Product Code. 18356686 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356686 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356686 Supplier Novus Biologicals Supplier No. NBP314676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Senataxin Monoclonal antibody specifically detects Senataxin in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Senataxin
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone BLR050F
Conjugate Unconjugated
Formulation Borate Buffered Saline (BBS) pH 8.2 with, BSA Free with 0.09% Sodium Azide
Gene Alias ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX
Host Species Rabbit
Immunogen between 900 and 950
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23064
Target Species Human
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.