missing translation for 'onlineSavingsMsg'
Learn More

Senataxin Antibody (4G1) - Azide and BSA Free, Novus Biologicals™

Product Code. 18338980 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338980 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338980 Supplier Novus Biologicals Supplier No. NBP280943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Senataxin Monoclonal specifically detects Senataxin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Senataxin
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 4G1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 5 μg/mL, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin : 1:200-1:500, Flow (Intracellular) : 5 μg/mL, CyTOF-ready
Formulation PBS
Gene Alias ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX
Gene Symbols SETX
Host Species Mouse
Immunogen Partial recombinant human Senataxin protein (between amino acids 2300-2600) [UniProt# Q7Z333]
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23064
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.