Learn More
SemperIt™ Semperguard™ Nitrile Xtra lite Protective Gloves
Product Code. p-8002734
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 180
Pack of 200
Product Code. Size unitSize
11807202 Large Pack of 200
11897192 Medium Pack of 200
11887192 Small Pack of 200
11817202 X-Large Pack of 180
4 options
Product Code. 11807202
Brand: SemperIt™ 3000001619
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from nitrile. Semperguard™ Nitrile Xtra lite Protective Gloves are disposable.
  • Nitrile
  • Powder free
  • Lavender blue
  • Disposable
  • Rolled Cuff Style
  • Ambidextrous
  • Latex Free
  • Tensile Strength of 14/14MPa
  • CE Class I, EN 420, EN 374, EN 455, PPE Category III certified 
Specifications
Certifications/Compliance CE Class I, EN 420, EN 374, EN 455, PPE Category III
Color Purple
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
Glove Cuff Style Rolled
Model 816780237
Tensile Strength 14 MPa
Length (Metric) 240 mm