missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Nitrile Xpert Powder Free Gloves

Product Code. 15523536
Click to view available options
Size:
Small
Large
X-Large
Unit Size:
Pack of 100
Pack of 90
This item is not returnable. View return policy

Product Code. 15523536

Brand: Semperit™ 3000001630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blue nitrile gloves are powder free and resistant to skin irritants.

  • PPE Category III complex risks
  • Excellent resistance to skin irritants
  • Suitable for food contact
  • Powder free
  • Chemical resistant
  • No silicone added

Specifications

Certifications/Compliance CE Class I; EN 420, EN 374, EN 455; PPE Category III
Classification Category III
Description Semperguard™ Nitrile Xpert Powder Free Gloves
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
Thickness (Metric) 0.26 mm
For Use With (Application) Pharmaceutical and laboratory applications
Glove Cuff Style Rolled
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.