missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ G01R Red Lightweight Latex Rubber Gloves

Product Code. 15690924 Shop All Ansell Products
Click to view available options
Size:
6.5
8.5
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 15690924

Brand: Ansell Edmont™ D013065

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Resistant to many cleaning chemicals used in laboratories. Ansell Edmont™ G01R Red Lightweight Latex Rubber Gloves are extra thick natural rubber latex gloves with a sand blast grip pattern, lightly flocked liner, and extended-length cuff with fluted shape.

Ideal for use in laboratories with short-term chemical contact, light duty maintenance, painting, printing, pharmaceutical production and applications requiring protection and sensitivity.
  • Resistant to many cleaning chemicals used in laboratories
  • Flexible and comfortable for excellent dexterity
  • Lightly flocked interior for better sensitivity
  • Sandblasted finish to guarantee a secure grip in dry or wet conditions
  • Extended-length, tear-resistant beaded cuff for extra protection and easy donning
  • Fluted cuff design enables ventilation to the hand
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN420 (General requirements), EN388 (Mechanical Hazards) and EN374 (Chemical and Microorganisms Hazards)
Classification Manufactured under EC quality assurance system carried out by the BSI.
Color Red
Exterior Finish Sand Blast
Hand Specific
Latex Free No
Material Rubber Latex
Size 6.5
Thickness (Metric) 0.70 mm
Glove Cuff Style Beaded, Fluted
Lining Lightly flocked
Packaging Type Bagged
Quantity 12 Pair
Resistant To Many laboratory cleaning chemicals
Series G01R
Length (Metric) 325 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.