missing translation for 'onlineSavingsMsg'
Learn More

SemperIt™ Semperguard™ Latex Innercoated (IC) Powder-Free Gloves

Product Code. 11733273
Click to view available options
:
Large (8-9)
This item is not returnable. View return policy

Product Code. 11733273

Brand: SemperIt™ 3000001292

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from high quality soft latex film. Semperit™ Semperguard™ Latex Innercoated (IC) Powder-Free Gloves are available in lightly powdered and powder-free styles.

  • Manufactured from high quality soft latex film and available in lightly powdered and powder-free styles
  • Lightly powdered or powder-free
  • Microroughened outer surface offeres better grip
  • Powder-free gloves have a synthetic inner coating for ease of donning
  • Ambidextrous 
  • Rolled cuff

Specifications

Certifications/Compliance CE Class I, EN 420, EN 374, EN 455, PPE Category III
Coating Material Smooth inside, textured outside
Color Natural
Disposable Single-use
Hand Ambidextrous
Material Rubber
Powdered Powder Free
Size Large
Sterility Non-sterile
Thickness (Metric) 0.24 mm
For Use With (Application) Suitable for food contact
Glove Cuff Style Rolled
Quantity 100/Pk.
Tensile Strength 18/14 MPa
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.