missing translation for 'onlineSavingsMsg'
Learn More

Semperit™ Semperguard™ Green Nitrile Disposable Gloves
GREENER_CHOICE

Product Code. 15940057
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 180
Pack of 200
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 15940057

Brand: Semperit™ 3000008213

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Developed with sustainability in mind due to the unique production process, the glove offers skin-friendly hand protection, reduces the risk of allergy and feels soft to the skin. Comfortable and strong tensile strength ensures stability and a pleasant wearing experience even over longer periods.

  • PPE Cat. III
  • AQL 1.5
  • Powder-free
  • Dispoable, non-sterile
  • Comfortable to wear, even over longer periods of time
  • Skin-friendly formulation: no chlorine, no natural rubber latex proteins, no rubber accelerators
  • Manufactured while saving on water and energy, and reducing CO2 emissions
  • Synthetic inner coating
  • Textured fingertips provide extra grip
  • Stability and pleasant wearing experience
  • Glove material is resistant to blood and other body fluids, and protects against bacteria, fungi and viruses including SARS-CoV-2, Hepatitis B, Hepatitis C, and HIV
  • Ideal for applications including Personal Protection, Food/Catering, Consumer/Retail, Beauty/Body Art and Hairdressing

Specifications

Certifications/Compliance (Europe) PPE Category III, EN 420, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN ISO 374-5, EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 2859-1, ASTM D6319, ASTM F1671
Chlorinated No
Accelerator Free Yes
Applications Industrial
AQL 1.5
Color Mint Green
Disposable Single-use
Exterior Finish Fingertip Textured
Material Nitrile
Powdered Powder Free
Size Small
Sterility Non-sterile
Glove Cuff Style Rolled
Series Semperguard™ Green
Tensile Strength 6 Newton
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.