missing translation for 'onlineSavingsMsg'
Learn More

Semicarbazide Hydrochloride, Extra Pure, SLR, Fisher Chemical

Product Code. 10325910 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10325910 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10325910 Supplier Fisher Chemical Supplier No. S/0560/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 563-41-7
Melting Point 170°C
Color White
pH 1.5
Packaging Plastic powder jar
Molecular Formula CH6ClN3O
MDL Number MFCD00013009
Quantity 100 g
Synonym semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
InChI Key XHQYBDSXTDXSHY-UHFFFAOYSA-N
SMILES [H+].[Cl-].NNC(N)=O
IUPAC Name aminourea;hydrochloride
Molecular Weight (g/mol) 111.53
PubChem CID 11236
ChEBI CHEBI:82532
Formula Weight 111.53g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Semicarbazide hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.