Learn More
Semicarbazide hydrochloride, 99%, Thermo Scientific Chemicals
Description
Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.
Solubility
Soluble in water (>100g/L).
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
Melting Point | ∼176°C (decomposition) |
pH | 1.5 |
Odor | Odorless |
Linear Formula | H2NCONHNH·HCl |
Quantity | 500 g |
UN Number | UN2811 |
Beilstein | 3593642 |
Merck Index | 14,8444 |
Solubility Information | Soluble in water (>100g/L). |
Formula Weight | 111.53 |
Show More |
RUO – Research Use Only