missing translation for 'onlineSavingsMsg'
Learn More

Semi-Volatile Calibration Standard, SPEX CertiPrep™

Product Code. 12797496 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12797496 1 mL 1mL
12707506 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12797496 Supplier SPEX Certiprep Supplier No. BIGBN2
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2,4-Trichlorobenzene120-82-12000μg/mL
1,2-Dichlorobenzene95-50-12000μg/mL
1,3-Dichlorobenzene541-73-12000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
2,4-Dinitrotoluene121-14-22000μg/mL
2,6-Dinitrotoluene606-20-22000μg/mL
2-Chloronaphthalene91-58-72000μg/mL
2-Methylnaphthalene91-57-62000μg/mL
2-Nitroaniline88-74-42000μg/mL
3-Nitroaniline99-09-22000μg/mL
4-Bromodiphenyl ether101-55-32000μg/mL
4-Chloroaniline (p-Chloroaniline)106-47-82000μg/mL
4-Chlorophenyl-phenyl ether7005-72-32000μg/mL
4-Nitroaniline100-01-62000μg/mL
Acenaphthene83-32-92000μg/mL
Acenaphthylene208-96-82000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(g,h,i)perylene191-24-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Benzyl alcohol100-51-62000μg/mL
Bis(2-chloro-1-methylethyl) ether108-60-12000μg/mL
bis(2-Chloroethoxy)methane111-91-12000μg/mL
bis(2-Chloroethyl)ether111-44-42000μg/mL
Bis(2-Ethylhexyl)phthalate117-81-72000μg/mL
Butylbenzyl phthalate85-68-72000μg/mL
Chrysene218-01-92000μg/mL
Dibenz(a,h)anthracene53-70-32000μg/mL
Dibenzofuran132-64-92000μg/mL
Diethyl phthalate84-66-22000μg/mL
Dimethyl phthalate131-11-32000μg/mL
Di-n-butyl phthalate84-74-22000μg/mL
Di-n-octyl phthalate117-84-02000μg/mL
Fluoranthene206-44-02000μg/mL
Fluorene86-73-72000μg/mL
Hexachlorobenzene118-74-12000μg/mL
Hexachlorobutadiene87-68-32000μg/mL
Hexachlorocyclopentadiene77-47-42000μg/mL
Hexachloroethane67-72-12000μg/mL
Indeno(1,2,3-c,d)pyrene193-39-52000μg/mL
Isophorone78-59-12000μg/mL
Naphthalene91-20-32000μg/mL
Nitrobenzene98-95-32000μg/mL
N-Nitrosodi-n-propylamine621-64-72000μg/mL
N-Nitrosodiphenylamine86-30-62000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Semi-volatile Calibration Standard
Packaging Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Methods 625, 8270, CLP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methylene Chloride/Benzene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Components in Methylene Chloride:Benzene at 2000μg/mL
Acenaphthene    Dimethyl Phthalate
Acenaphthylene    Di-n-butyl Phthalate
Anthracene    2,4-Dinitrotoluene
Benszo(a)anthracene    2,6-Dinitrotoluene
Benzo(a)pyrene    Di-n-octyl Phthalate
Benzo(b)fluoranthene    Fluoranthene
Benzo(g,h,i)perylene    Fluorene
Benzo(k)fluoranthene    Hexachlorobenzene
Benzyl Alcohol    Hexachlorobutadiene
bis(2-chloroethoxy)methane    Hexachlorocyclopentadiene
bis(2-chloroethyl)ether    Hexachloroethane
bis(2-chloroisopropyl)ether    Indeno(1,2,3-cd)pyrene
bis(2-ethylhexyl)phthalate    Isophorone
4-Bromophenyl phenyl ether    2-Methylnaphthalene
Butylbenzyl phthalate    Naphthalene
4-chloroaniline    2-Nitroaniline
2-chloronaphthalene    3-Nitroaniline
4-chlorophenyl-phenyl ether    4-Nitroaniline
Chrysene    Nitrobenzene
Dibenz(a,h)anthracene    N-Nitroso-di-n-propylamine
Dibenzofuran    N-Nitrosodiphenylamine
1,2-Dichlorobenzene    Phenanthrene
1,3-Dichlorobenzene    Pyrene
1,4-Dichlorobenzene    1,2,4-Trichlorobenzene
Diethyl Phthalate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Semi-volatile Calibration Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: anthracene, hexachlorobuta-1,3-diene, 1,2,4-trichlorobenzene

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.