missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Semi-Dry Blotters

Product Code. p-8272077
Click to view available options
Dimensions (L x W x H):
7.5 x 17 x 17 cm
7.5 x 27 x 27 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12388556

Brand: Fisherbrand™ V10SDB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for fast transfer of proteins and nucleic acids without the need for gel cassettes and tanks.

  • Designed to produce an extremely uniform system for the transfer of proteins and nucleic acids.
  • Very low buffer volume and current, 0.8mA per cm2 of gel, produces very little heat, preventing band distortion
  • Transfers can be achieved in less than one hour, depending on the molecular weight of the molecules
  • Either single gels or multiple gel stacks can be used with the units
  • Colour coded electrodes: One electrode is manufactured from stainless steel and the other from platinised titanium, making them virtually corrosion free
  • Fast transfer times
  • Available in two sizes
 V10-SDB: 10 x 10cm format - accommodates one 10 x 10cm mini-gel
 V20-SDB: 20 x 20cm format - accommodates up to four 10 x 10cm mini-gels or one 20 x 20cm maxi-gel

Specifications

Type Semi Dry Electroblotting System
Gel Dimensions Active Transfer Area: L x W: 10 x 10 cm
Dimensions (L x W x H) 7.5 x 17 x 17 cm
Electrical Requirements Recommended Power Supplies: EV243
Amperage 80 mA / 0.8 mAcm-2
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato