missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semenogelin II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Semenogelin II |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Semenogelin II Polyclonal specifically detects Semenogelin II in Human samples. It is validated for Western Blot.Specifications
| Semenogelin II | |
| Polyclonal | |
| Rabbit | |
| NP_002999 | |
| 6407 | |
| Synthetic peptide directed towards the N terminal of human SEMG2. Peptide sequence GQKGQHYFGQKDQQHTKSKGSFSIQHTYHVDINDHDWTRKSQQYDLNALH. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| semenogelin IISGIISemenogelin 2, semenogelin-2 | |
| SEMG2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title