missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 7A Antibody (MM0537-99D22), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11884R
This item is not returnable.
View return policy
Description
Semaphorin 7A Monoclonal specifically detects Semaphorin 7A in Human samples. It is validated for Western Blot.
Specifications
| Semaphorin 7A | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SEMA7A | |
| Human recombinant Semaphorin 7A (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0537-99D22 | |
| Western Blot | |
| CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, SEMAK1, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L | |
| Rat | |
| Protein G purified | |
| Adaptive Immunity, Immunology, Neuroscience | |
| 8482 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction