missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Semaphorin 4D/CD100 Antibody (pepinemab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
£157.00 - £695.00
Specifications
| Antigen | Semaphorin 4D/CD100 |
|---|---|
| Clone | pepinemab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045720
|
Novus Biologicals™
NBP3-28669-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047721
|
Novus Biologicals™
NBP3-28669-50ug |
50 μg | |||||||
|
30048526
|
Novus Biologicals™
NBP3-28669-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Semaphorin 4D/CD100 Monoclonal antibody specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| Semaphorin 4D/CD100 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis, Immunology | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| SEMA4D/ CD100 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| pepinemab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q92854 | |
| 10507 | |
| IgG4 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title