missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Semaphorin 4D/CD100 Antibody Pair [HRP]

Product Code. p-7183135 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18279857

Brand: Novus Biologicals™ NBP27959715PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Semaphorin 4D/CD100.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 47 pg/mL
Assay Sensitivity 46.88-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Semaphorin 4D/CD100.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Gene ID (Entrez) 10507
Gene Symbol SEMA4D
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL rabbit anti-Semaphorin 4D/CD100 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse Semaphorin 4D/CD100 monoclonal antibody.
  • Standard: Each vial contains 150 ng of recombinant mouse Semaphorin 4D/CD100.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Immunology
    Storage Requirements Storage is content dependent.
    Target Semaphorin 4D/CD100
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.