missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (JE63-21), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32119
This item is not returnable.
View return policy
Description
Semaphorin 4D/CD100 Monoclonal antibody specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Western Blot
Specifications
| Semaphorin 4D/CD100 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| Synthetic peptide within human Semaphorin 4D/CD100 aa 126-175/862. (Uniprot: Q92854) | |
| 100 μL | |
| Apoptosis, Immunology | |
| 10507 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE63-21 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction