missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 3C Antibody (RM0315-7F18), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11877AF750
This item is not returnable.
View return policy
Description
Semaphorin 3C Monoclonal specifically detects Semaphorin 3C in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Semaphorin 3C | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SEMA3C | |
| Mouse recombinant Semaphorin 3C | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0315-7F18 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| (semaphorin) 3C, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema E, SEMAE, semaphorin E, semaphorin-3C, Semaphorin-E, SemE | |
| Rat | |
| Protein A or G purified | |
| Neuroscience | |
| 10512 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction