missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 3B Antibody, Novus Biologicals™

Product Code. 18368500 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368500 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368500 Supplier Novus Biologicals Supplier No. NBP286801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Semaphorin 3B Polyclonal specifically detects Semaphorin 3B in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Semaphorin 3B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias (semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, semaV
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of Human Semaphorin 3B
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7869
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.