missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 3B Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2218R
This item is not returnable.
View return policy
Description
Semaphorin 3B Polyclonal specifically detects Semaphorin 3B in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Semaphorin 3B | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q13214 | |
| SEMA3B | |
| A synthetic peptide made to an internal portion of the human SEMA3B protein sequence (between residues 100-200). [UniProt# Q13214] | |
| 0.05 mL | |
| Cancer, Growth and Development, Neuronal Cell Markers | |
| 7869 | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| (semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, semaV | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, human, rat, dog, bovine, and primate. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction