missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 3A Antibody (1040525) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB109031JF635
This item is not returnable.
View return policy
Description
Semaphorin 3A Monoclonal antibody specifically detects Semaphorin 3A in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| Semaphorin 3A | |
| Monoclonal | |
| Janelia Fluor 635 | |
| (semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD | |
| Chinese Hamster Ovary cell line CHO-derived mouse Semaphorin 3A protein, Asn21-Lys747, Accession # O08665 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 1040525 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Neuroscience | |
| 10371 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction