missing translation for 'onlineSavingsMsg'
Learn More

Semaglutide TFA, MedChemExpress

Product Code. 16497527
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497527 5 mg 5mg
16421209 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497527 Supplier Medchem Express Supplier No. HY114118A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Semaglutide TFA, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide TFA has the potential for type 2 diabetes treatment.

Semaglutide TFA, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide TFA has the potential for type 2 diabetes treatment.

Specifications

Chemical Name or Material Semaglutide TFA
Physical Form Solid
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C189H292F3N45O61
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 100 mg/mL (23.65 mM; Need ultrasonic)
SMILES [Semaglutide (TFA salt)]
Molecular Weight (g/mol) 4227.66
Formula Weight 4227.66
Percent Purity 98.26%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.