missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaglutide, MedChemExpress
Description
Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment.
Specifications
Specifications
| Chemical Name or Material | Semaglutide |
| Physical Form | Solid |
| CAS | 910463-68-2 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C187H291N45O59 |
| For Use With (Application) | Metabolism-sugar/lipid metabolism |
| Solubility Information | DMSO : 33.33 mg/mL (8.10 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (3.04 mM; ultrasonic and adjust pH to 2 with HCl) |
| SMILES | [Semaglutide] |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?