Learn More
Invitrogen™ SEMA7A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588314
Description
Immunogen sequence: QGHLRSGPRI FAVWKGHVGQ DRVDFGQTEP HTVLFHEPGS SSVWVGGRGK VYLFDFPEGK NASVRTVNIG STKGSCLDKR DCENYITLLE RRSEGLLACG TNARHPSCWN LVNGTVVPLG EMRGYAPFSP DENSLVLFEG DEVYSTIRKQ EYNGKIPRFR RIRGESELYT SDTVMQNPQF IKATIVHQDQ AYDDKIYYFF REDNPDKNPE APLNVSRVAQ LCRGDQGGES SLSVSK; Positive Samples: A549, HT-29, Raji; Cellular Location: Cell membrane, Extracellular side, GPI-anchor, Lipid-anchor.
CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
Specifications
| SEMA7A | |
| Polyclonal | |
| Unconjugated | |
| SEMA7A | |
| 2900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8482 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.4 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75326 | |
| SEMA7A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 45-280 of human SEMA7A (NP_0036031). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.