Learn More
Invitrogen™ SEMA7A Monoclonal Antibody (MEM-150)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119203
Description
This antibody reacts with CD108 (JMH blood group antigen), a 80 kDa GPI-anchored extracellular glycoprotein expressed on various cell types including erythrocytes, lymphoblasts; at low levels it is present on circulating lymphocytes. Western Blot: non-reducing conditions.
CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
Specifications
| SEMA7A | |
| Monoclonal | |
| 1 mg/mL | |
| TBS with 15 mM sodium azide; pH 8 | |
| O75326 | |
| SEMA7A | |
| T cell line HPB-ALL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| MEM-150 | |
| Unconjugated | |
| SEMA7A | |
| 2900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk1 | |
| Mouse | |
| Sequential Chromatography | |
| RUO | |
| 8482 | |
| 4°C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.