missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA6B, Mouse anti-Human, Clone: 2H7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010501-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: PEEPPPLSVAPRDYLNHYPVFVGSGPGRLTPAEGADDLNIQRVLRVNRTLFIGDRDNLYRVELEPPTSTELRYQRKLTWRSNPSDINVCRMKGKQEGECSpecifications
| SEMA6B | |
| Monoclonal | |
| Unconjugated | |
| sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B | |
| SEM-SEMA-Y/SEMA-VIB/SEMAN/semaZ | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2H7 | |
| In 1x PBS, pH 7.4 | |
| NM_020241 | |
| SEMA6B | |
| SEMA6B (NP_064626, 28 a.a. ∼ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10501 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction