missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA6A Recombinant Rabbit Monoclonal Antibody (HL2120)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550386
This item is not returnable.
View return policy
Description
SEMA6A Recombinant Monoclonal Antibody for Western Blot
SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
Specifications
| SEMA6A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O35464, Q9H2E6 | |
| SEMA6A | |
| Recombinant fragment of human SEMA6A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2120 | |
| Unconjugated | |
| SEMA6A | |
| 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20358, 361324, 57556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction