missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£161.00 - £380.00
Specifications
| Antigen | SEMA5B |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17940093
|
Invitrogen™
PA5145463 |
200 μL |
£380.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17930863
|
Invitrogen™
PA5145464 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May act as positive axonal guidance cues.Specifications
| SEMA5B | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Pig, Rat | |
| Q60519, Q9P283 | |
| 100519925, 20357, 303901, 54437 | |
| Recombinant Human SEMA5B protein, Leu36-Pro602 (Accession #Q9P283). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AI893641; Kiaa1445; mKIAA1445; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B; sema G; Sema5b; Semag; semaphorin 5B; semaphorin-5B; Semaphorin-G; SemG; UNQ5867/PRO34001 | |
| SEMA5B | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| SEMA5B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title