missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA5B Monoclonal Antibody (C3), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £408.00
Specifications
| Antigen | SEMA5B |
|---|---|
| Clone | C3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17654046
|
Invitrogen™
MA543589 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17644046
![]() |
Invitrogen™
MA543588 |
100 μL |
£408.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May act as positive axonal guidance cues.Specifications
| SEMA5B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| AI893641; Kiaa1445; mKIAA1445; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B; sema G; Sema5b; Semag; semaphorin 5B; semaphorin-5B; Semaphorin-G; SemG; UNQ5867/PRO34001 | |
| SEMA5B | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9P283 | |
| 54437 | |
| Gly350-Pro602 (Accession Q9P283), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SEMA5B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title