missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110683
This item is not returnable.
View return policy
Description
Immunogen sequence: PWTKCSATCG GGHYMRTRSC SNPAPAYGGD ICLGLHTEEA LCNTQPCPES WSEWSDWSEC EASGVQVRAR QCILLFPMGS QCSGNTTESR PCVFDSNFIP EVSVARSSSV EEKRCGE Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 94%.
Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).
Specifications
| SEMA5A | |
| Polyclonal | |
| Unconjugated | |
| SEMA5A | |
| 5930434A13; 9130201M22Rik; AI464145; Ensembl:ENSRNOP00000016506}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; sema F; SEMA5A; sema5a {ECO:0000312; SEMAF; semaphorin 5A; semaphorin F; semaphorin-5A; semaphorin-5A {ECO:0000250; Semaphorin-F; semF; UniProtKB:Q13591} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9037 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13591 | |
| SEMA5A | |
| Recombinant Protein corresonding to Human SEMA5A. Recombinant protein control fragment (Product #RP-100996). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction