missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA4F Monoclonal Antibody (780225)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561781
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.
Specifications
| SEMA4F | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Z123 | |
| SEMA4F | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sema-4F, Arg41-Gly667. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 780225 | |
| Unconjugated | |
| SEMA4F | |
| M-SEMA; m-Sema-M; PRO2353; S4F; sema domain immunoglobulin domain transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F; sema domain, immunoglobulin domain (Ig), TM domain, and short cy; sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4F; Sema M; Sema W; Sema4f; SEMAM; semaphorin 4f; semaphorin M; semaphorin W; semaphorin-4F; Semaphorin-M; semaphorin-W; SEMAW; ssemaphorin 4F | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction