Learn More
Invitrogen™ SEMA4C Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547814
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) Sema4C is observed, and approximately 10% cross-reactivity with recombinant human (rh) Sema4G is observed, and less than 5% cross-reactivity with rhSema3B and rmSema3F is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
SEMA4C (semaphorin-4C), also known as SEMAF, is an 833 amino acid single-pass type I membrane protein that contains one sema domain, one PSI domain and one Ig-like C2-type domain. Expressed in a variety of tissues, including lung, kidney and brain, SEMA4C interacts with GIPC and NCDN and is thought to play a role in the formation of neural networks during development.
Specifications
| SEMA4C | |
| Polyclonal | |
| Unconjugated | |
| SEMA4C | |
| AI426163; FLJ20369; KIAA1739; MGC126382; MGC126383; M-Sema F; M-SEMA-F; RGD1562837; S4c; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Sema I; SEMA4C; SEMACL1; SEMAF; SEMAI; semaphorin 4C; semaphorin-4C; Semaphorin-C-like 1; UNQ5855/PRO34487 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 54910 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9C0C4 | |
| SEMA4C | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4C Ala21-Gly663. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.