missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA4C Polyclonal Antibody

Description
Positive Samples: MCF7, BT-474, Mouse heart; Cellular Location: Cell junction, Cytoplasmic vesicle, Single-pass type I membrane protein, postsynaptic cell membrane, postsynaptic density, secretory vesicle, synapse, synaptic vesicle membrane.
SEMA4C (semaphorin-4C), also known as SEMAF, is an 833 amino acid single-pass type I membrane protein that contains one sema domain, one PSI domain and one Ig-like C2-type domain. Expressed in a variety of tissues, including lung, kidney and brain, SEMA4C interacts with GIPC and NCDN and is thought to play a role in the formation of neural networks during development.
Specifications
Specifications
| Antigen | SEMA4C |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SEMA4C |
| Gene Accession No. | Q64151, Q9C0C4 |
| Gene Alias | AI426163; FLJ20369; KIAA1739; MGC126382; MGC126383; M-Sema F; M-SEMA-F; RGD1562837; S4c; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Sema I; SEMA4C; SEMACL1; SEMAF; SEMAI; semaphorin 4C; semaphorin-4C; Semaphorin-C-like 1; UNQ5855/PRO34487 |
| Gene Symbols | SEMA4C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?