missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEMA4C Monoclonal Antibody (660016)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561630

Product Code. 30535114

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

SEMA4C (semaphorin-4C), also known as SEMAF, is an 833 amino acid single-pass type I membrane protein that contains one sema domain, one PSI domain and one Ig-like C2-type domain. Expressed in a variety of tissues, including lung, kidney and brain, SEMA4C interacts with GIPC and NCDN and is thought to play a role in the formation of neural networks during development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEMA4C
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9C0C4
SEMA4C
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4C, Ala21-Gly663 (predicted).
100 μg
Primary
Human
Antibody
IgG1
Western Blot
660016
Unconjugated
SEMA4C
AI426163; FLJ20369; KIAA1739; MGC126382; MGC126383; M-Sema F; M-SEMA-F; RGD1562837; S4c; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Sema I; SEMA4C; SEMACL1; SEMAF; SEMAI; semaphorin 4C; semaphorin-4C; Semaphorin-C-like 1; UNQ5855/PRO34487
Mouse
Protein A/G
RUO
54910
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.