Learn More
SEMA4B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29191-1-AP-150UL
This item is not returnable.
View return policy
Description
The SEMA4B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SEMA4B. This antibody recognizes human antigen. The SEMA4B antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| SEMA4B | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| KIAA1745, SEMA4B, SEMAC, SemC | |
| 150 μL | |
| 10509 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |