missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA4B, Mouse anti-Human, Clone: 4B2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010509-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: GEKPCEQVQFQPNTVNTLACPLLSNLATRLWLRNGAPVNASASCHVLPTGDLLLVGTQQLGEFQCWSLEEGFQQLVASYCPEVVEDGVADQTDEGGSVSpecifications
| SEMA4B | |
| Monoclonal | |
| Unconjugated | |
| sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B | |
| KIAA1745/MGC131831/SEMAC/SemC | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4B2 | |
| In 1x PBS, pH 7.4 | |
| NM_020210 | |
| SEMA4B | |
| SEMA4B (NP_064595, 592 a.a. ∼ 689 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10509 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction