missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA3A Monoclonal Antibody (M317)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547732
This item is not returnable.
View return policy
Description
The antibody detects a 95kDa* protein corresponding to the apparent molecular mass of Sema3A on SDS-PAGE immunoblots of human recombinant Sema3A and adult mouse brain lysates.
This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease.
Specifications
| SEMA3A | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| O08665, Q14563, Q63548 | |
| SEMA3A | |
| Clone M317 was generated from a synthetic peptide (coupled to carrier protein) corresponding to amino acids in the central region of human Sema3A. The sequence used is highly conserved in rat and mouse Sema3A, and has low homology to other semaphorin family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| M317 | |
| Unconjugated | |
| SEMA3A | |
| COLL1; coll-1; collapsin 1; collapsin-1; HH16; Hsema-I; Hsema-III; MGC133243; sema D; sema domain, immunoglobulin domain (Ig), short basic domain, sec; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; sema III; SEMA1; SEMA3A; Semad; SEMAIII; SEMAL; Semaphorin 3a; semaphorin D; semaphorin III; semaphorin L; semaphorin-3A; Semaphorin-D; SemD | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 10371, 20346, 29751 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction