missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA3A Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543592
This item is not returnable.
View return policy
Description
This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease.
Specifications
| SEMA3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q14563 | |
| SEMA3A | |
| Pro580-Thr664, with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C2 | |
| Unconjugated | |
| SEMA3A | |
| COLL1; coll-1; collapsin 1; collapsin-1; HH16; Hsema-I; Hsema-III; MGC133243; sema D; sema domain, immunoglobulin domain (Ig), short basic domain, sec; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; sema III; SEMA1; SEMA3A; Semad; SEMAIII; SEMAL; Semaphorin 3a; semaphorin D; semaphorin III; semaphorin L; semaphorin-3A; Semaphorin-D; SemD | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction