missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SELS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543131
This item is not returnable.
View return policy
Description
Peptide sequence: PDVVVKRQEA LAAARLKMQE ELNAQVEKHK EKLKQLEEEK RRQKIEMWDS Sequence homology: Cow: 93%; Dog: 86%; Goat: 75%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 92%; Rat: 93%; Yeast: 90%; Zebrafish: 100%.
This gene encodes a transmembrane protein that is localized in the endoplasmic reticulum (ER). It is involved in the degradation process of misfolded proteins in the ER, and may also have a role in inflammation control. It participates in the transfer of misfolded proteins from the ER to the cytosol, where they are destroyed by the proteasome in a ubiquitin-dependent manner. This protein is a selenoprotein, containing the rare amino acid selenocysteine (Sec). Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal.
Specifications
| SELS | |
| Polyclonal | |
| Unconjugated | |
| SELENOS | |
| 1500011E07Rik; AD-015; ADO15; C78786; chondroitin sulfate synthase 1; CHSY1; H4; H-4; H47; H-47; histocompatibility 47; MGC104346; MGC2553; minor histocompatibility antigen H47; SBBI8; selenoprotein S; Selenos; SELS; SEPS1; sg2; similar to selenoprotein S; Tanis; valosin-containing protein-interacting membrane protein; VCP interacting membrane selenoprotein; VCP-interacting membrane protein; VCP-interacting membrane selenoprotein; vimp; wu:fc16a03; zgc:136970 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BQE4 | |
| SELENOS | |
| Synthetic peptide directed towards the middle region of human SELS. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction