missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELS Mouse anti-Human, Clone:7F8G1, Abnova™
Mouse monoclonal antibody raised against recombinant human SELS.
Brand: Abnova MAB17233.100ug
This item is not returnable.
View return policy
Description
This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3′ UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response. Two alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]Specifications
| SELS | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
| SELS | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 7F8G1 | |
| selenoprotein S | |
| In PBS (0.05% sodium azide) | |
| AD-015, ADO15, MGC104346, MGC2553, SBBI8, SEPS1, VIMP | |
| Recombinant protein corresponding to amino acid 1-187 of human SELS from E. coli. | |
| 55829 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction