missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELS Antibody (7F8G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61876-0.025ml
This item is not returnable.
View return policy
Description
SELS Monoclonal specifically detects SELS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SELS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| ADO15, MGC104346, SBBI8, selenoprotein S, SelS, SEPS1, VCP-interacting membrane protein, VIMPMGC2553 | |
| Mouse | |
| 21.2 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 7F8G1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| VIMP | |
| Purified recombinant fragment of human VIMP (AA: 1-187) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 55829 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction