missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELM Polyclonal Antibody
SELM Polyclonal Antibody
Brand: Invitrogen PA572639
This item is not returnable.
View return policy
Description
SELM Polyclonal Antibody for Western Blot
Specifications
| SELM | |
| Polyclonal | |
| Unconjugated | |
| SELENOM | |
| cb693; fc13f12; SELENOM; Selenoprotein M; selenoprotein M precursor; selenoprotein SelM; selm; SelM protein; sepm; sepm protein; wu:fc13f12; zgc:103431 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 352913 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q802G7 | |
| SELENOM | |
| KLH conjugated synthetic peptide between 96-119 amino acids from the C-terminal region of DANRE selm. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction