missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SELM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550260
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SELM protein.
A growing body of evidence relates selenium to cancer prevention, immune system function, male fertility, cardiovascular disorder, control of the aging and neurodiseases process. Selenoproteins are thought to be responsible for the majority of these biomedical effects of selenium. Approximately 17 selenoproteins have been identified until now. Although the function of many selenoproteins are unknown, some play important roles in anti-oxidant mechanisms. It has been also implicated in the regulation of signaling pathways through catalysis of thiol/disulfide exchange. Selenoprotein M (SelM) is especially expressed in a mammalian brain and is localized to the perinuclear structures (Golgi/ER). The roles of selM have not been clearly identified until present time.
Specifications
| SELM | |
| Polyclonal | |
| Unconjugated | |
| SELENOM | |
| cb693; fc13f12; SELENOM; Selenoprotein M; selenoprotein M precursor; selenoprotein SelM; selm; SelM protein; sepm; sepm protein; wu:fc13f12; zgc:103431 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140606 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8WWX9 | |
| SELENOM | |
| Fusion protein corresponding to residues near the C terminal of human elenoprotein M. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction