missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yes 2 Solutions™ Sellotape double-sided tape, 25mm x 33m
Description
Versatile tape coated with strong adhesive on both sides to create an instant dry adhesive bond. Easy tear and easily removable backing paper for effortless use. Ideal for display, mounting and exhibition work.
- Size: 25mm x 33m
- Pack of six rolls
Specifications
Specifications
| Length (Metric) | 33 m |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?