Learn More
Globe Scientific Self-Standing Transport Tubes
Product Code. 31923153 Shop All Globe Scientific Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
Color:
Amber
Clear
Closure Included:
No
Yes
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Closure Included Unit Size
31923153 5 mL Amber Yes Pack of 1000
31923634 5 mL Clear No Pack of 1000
31921775 10 mL Clear Yes Pack of 1000
31922351 10 mL Clear No Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31923153 Supplier Globe Scientific Supplier No. 6101AM
Upgrade Your Laboratory Essentials with Globe Scientific
Use promo code 27959
View all promotions
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for the safe storage and transport of biological materials for 6101 or 6102 series transport tubes
  • The transparent polypropylene (PP) vials allow unobstructed viewing of the sample
  • The vials are shatter-proof and may be exposed to freezing temperatures
  • They feature leak-resistant screw caps that can be removed or sealed in a single turn
Specifications
Color Amber
Autoclavable Not Autoclavable
Capacity (Metric) 5 mL
Bottom Shape Conical (Self-standing)
Material Polyethylene
Thread Style External
Sterility Non-sterile
Graduated Yes
Closure Included Yes
Product Type Transport Tube (Self-Standing)